(+)-Dimethyl-L-tartrate ≥98.0%
Supplier: TCI America
Synonyms:
(+)-Dimethyl L-tartrate
CAS Number: 608-68-4
MDL Number: MFCD00064437
Molecular Formula: C6H10O6
Molecular Weight: 178.14
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 118
Melting point (°C): 62
Specific rotation [a]20/D: 21 deg (C=1, H2O)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(+)-Tartaric Acid Dimethyl Ester
Formula:
C₆H₁₀O₆ MW: 178.14 g/mol Boiling Pt: 118 °C Melting Pt: 62 °C Density: 1.3403 g/cm³ (15 °C) |
MDL Number:
MFCD00064437 CAS Number: 608-68-4 EINECS: 210-166-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...