Diethyl suberate ≥98.0%
Supplier: TCI America
Synonyms:
Suberic acid diethyl ester
, Diethyl octanedioate
, 1,8-Octanedioic acid diethyl ester
CAS Number: 2050-23-9
MDL Number: MFCD00009217
Molecular Formula: C12H22O4
Molecular Weight: 230.30
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 283
Flash Point (°C): 110
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: Suberic Acid Diethyl Ester, Octanedioic Acid Diethyl Ester, Diethyl Octanedioate
Formula:
C₁₂H₂₂O₄ MW: 230.3 g/mol Boiling Pt: 283 °C Melting Pt: 5 °C Density: 0.98 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00009217 CAS Number: 2050-23-9 EINECS: 218-084-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...