D-(+)-Sorbose ≥98.0% (by HPLC)
Supplier: TCI America
S0390-100MG
TCS0390-100MGEA
82.22
USD
TCS0390-100MG
D-(+)-Sorbose ≥98.0% (by HPLC)
D(+)-Sorbose
CAS Number: 3615-56-3
MDL Number: MFCD00151095
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 158
Specific rotation [a]20/D: 40 deg (C=1, H2O)
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Boiling Pt: 380 °C (1013 hPa) Melting Pt: 158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151095 CAS Number: 3615-56-3 EINECS: 222-796-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...