1-Octadecene ≥99.5%

Supplier: TCI America

S0348-1ML
TCS0348-001MLEA 87.09 USD
TCS0348-001ML
1-Octadecene ≥99.5%
1-Octadecene

[Standard Material for GC]
CAS Number: 112-88-9
MDL Number: MFCD00009003
Molecular Formula: C18H36
Molecular Weight: 252.49
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 179
Flash Point (°C): 149
Freezing point (°C): 16
Specific Gravity (20/20): 0.79

Formula: C₁₈H₃₆
MW: 252.48 g/mol
Boiling Pt: 179 °C
Melting Pt: 17.5 °C
Density: 0.79 (20 °C)
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00009003
CAS Number: 112-88-9
EINECS: 204-012-9

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR