1-Dodecene ≥99.5%
Supplier: TCI America
Synonyms:
Dodec-1-ene
S0342-5ML
TCS0342-005MLEA
39.45
USD
TCS0342-005ML
1-Dodecene ≥99.5%
1-Dodecene
[Standard Material for GC]
CAS Number: 112-41-4
MDL Number: MFCD00008961
Molecular Formula: C12H24
Molecular Weight: 168.32
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 213
Flash Point (°C): 76
Specific Gravity (20/20): 0.76
Formula:
C₁₂H₂₄ MW: 168.32 g/mol Boiling Pt: 213 °C Melting Pt: –35 °C Density: 0.76 (20 °C) Flash Pt: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008961 CAS Number: 112-41-4 EINECS: 203-968-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...