Methyl octanoate ≥99.5%
Supplier: TCI America
Synonyms:
Caprylic acid methyl ester
[Standard Material for GC]
CAS Number: 111-11-5
MDL Number: MFCD00009551
Molecular Formula: C9H18O2
Molecular Weight: 158.24
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 193
Flash Point (°C): 76
Specific Gravity (20/20): 0.88
Product Categories: Analytical Science, Fatty Acid Methyl Esters (GC Standard)
Other Product Names / Synonyms: n-Octanoic Acid Methyl Ester
Formula:
C₉H₁₈O₂ MW: 158.24 g/mol Boiling Pt: 193 °C Melting Pt: –36.7 °C Density: 0.88 (20 °C) Flash Pt: 76 °C |
MDL Number:
MFCD00009551 CAS Number: 111-11-5 EINECS: 203-835-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...