n-Tetracosane ≥99.0%

Supplier: TCI America

Synonyms: N-tetracosane

S0296-1G
TCS0296-001GEA 36.84 USD
TCS0296-001G
n-Tetracosane ≥99.0%
n-Tetracosane

[Standard Material for GC]
CAS Number: 646-31-1
MDL Number: MFCD00009352
Molecular Formula: C24H50
Molecular Weight: 338.66
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 250
Melting point (°C): 51
Flash Point (°C): 194

Formula: H₃C(CH₂)₂₂CH₃
MW: 338.66 g/mol
Boiling Pt: 250 °C
Melting Pt: 51 °C
Density: 0.7787 g/cm³ (20 °C)
Flash Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00009352
CAS Number: 646-31-1
EINECS: 211-474-5
https://us.vwr.com/stibo/web/std.lang.all/40/37/28994037.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR