n-Heneicosane ≥99.5%

Supplier: TCI America

S0293-1G
TCS0293-001GEA 39.59 USD
TCS0293-001G
n-Heneicosane ≥99.5%
n-Heneicosane

[Standard Material for GC]
CAS Number: 629-94-7
MDL Number: MFCD00009346
Molecular Formula: C21H44
Molecular Weight: 296.58
Purity/Analysis Method: 99.5% (GC)
Form: Crystal
Color: White
Boiling point (°C): 356
Melting point (°C): 42
Flash Point (°C): 110


Product Categories: Analytical Science, n-Paraffins (GC Standard)
Other Product Names / Synonyms: Henicosane

Formula: H₃C(CH₂)₁₉CH₃
MW: 296.58 g/mol
Boiling Pt: 356 °C
Melting Pt: 42 °C
Density: 0.7919 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009346
CAS Number: 629-94-7
EINECS: 211-118-9
https://us.vwr.com/stibo/web/std.lang.all/40/34/28994034.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR