Sulfathiazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-N-(thiazol-2-yl)benzenesulphonamide4-Amino-N-(thiazol-2-yl)benzenesulfonamideN1-2-ThiazolylsulphanilamidN1-2-Thiazolylsulfanilamid

S0272-100G S0272-500G S0272-25G
TCS0272-100GEA 197.8 USD
TCS0272-100G TCS0272-500G TCS0272-025G
Sulfathiazole ≥98.0% (by HPLC, titration analysis)
Sulfathiazole

CAS Number: 72-14-0
MDL Number: MFCD00005319
Molecular Formula: C9H9N3O2S2
Molecular Weight: 255.31
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 200


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 2-(4-Aminobenzenesulfonamido)thiazole, 2-Sulfanilamidothiazole, N-2-Thiazolylsulfanilamide

Formula: C₉H₉N₃O₂S₂
MW: 255.32 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00005319
CAS Number: 72-14-0
EINECS: 200-771-5
https://us.vwr.com/stibo/web/std.lang.all/40/15/28994015.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR