Stearic acid hydrazide ≥95.0% (by GC, titration analysis)

Supplier: TCI America

S0222-25G
TCS0222-025GEA 57 USD
TCS0222-025G
Stearic acid hydrazide ≥95.0% (by GC, titration analysis)
Stearic acid hydrazide

CAS Number: 4130-54-5
MDL Number: MFCD00014764
Molecular Formula: C18H38N2O
Molecular Weight: 298.52
Purity/Analysis Method: 95.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 116

Formula: C₁₈H₃₈N₂O
Melting Pt: 116 °C
MDL Number: MFCD00014764
CAS Number: 4130-54-5
EINECS: 223-946-8
https://us.vwr.com/stibo/web/std.lang.all/39/89/28993989.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR