Ethyl piperazine-1-carboxylate ≥96.0%
Supplier: TCI America
Synonyms:
1-Ethoxycarbonyl piperazine
, Ethyl N-piperazinecarboxylate
CAS Number: 120-43-4
MDL Number: MFCD00005964
Molecular Formula: C7H14N2O2
Molecular Weight: 158.20
Purity/Analysis Method:
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 237
Flash Point (°C): 113
Specific Gravity (20/20): 1.09
Other Product Names / Synonyms: 1-Ethoxycarbonylpiperazine, 1-Piperazinecarboxylic Acid Ethyl Ester
Formula:
C₇H₁₄N₂O₂ MW: 158.2 g/mol Boiling Pt: 237 °C Density: 1.09 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00005964 CAS Number: 120-43-4 EINECS: 204-395-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...