Propagyl bromide ∼9.2 mol/L in toluene ≥80% stabilized
Supplier: TCI America
Synonyms:
3-Bromopropyne
P1272-500G
P1272-100G
P1272-25G
TCP1272-100GEA
117.1
USD
TCP1272-100G
TCP1272-25G
TCP1272-500G
Propagyl bromide ∼9.2 mol/L in toluene ≥80% stabilized
Propagyl bromide
(stabilized with MgO)
CAS Number: 106-96-7
MDL Number: MFCD00000241
Molecular Formula: C3H3Br
Molecular Weight: 118.96
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 18
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 3-Bromo-1-propyne
Formula:
C₃H₃Br Flash Pt: 18 °C |
MDL Number:
MFCD00000241 CAS Number: 106-96-7 EINECS: 203-447-1 UN: 2345 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...