4-Penten-1-ol ≥98.0%

Supplier: TCI America

Synonyms: 1-Penten-5-ol

P1243-25ML P1243-250ML
TCP1243-025MLEA 158.29 USD
TCP1243-025ML TCP1243-250ML
4-Penten-1-ol ≥98.0%
4-Penten-1-ol

CAS Number: 821-09-0
MDL Number: MFCD00002975
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 138
Flash Point (°C): 48
Specific Gravity (20/20): 0.85

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 138 °C
Density: 0.85 (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00002975
CAS Number: 821-09-0
EINECS: 212-473-2
UN: 1987
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/18/76/28991876.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR