Ethyl picolinate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl 2-pyridinecarboxylate
, Ethyl 2-picolinate
P1026-25G
TCP1026-025GEA
57.86
USD
TCP1026-025G
Ethyl picolinate ≥98.0%
Ethyl picolinate
CAS Number: 2524-52-9
MDL Number: MFCD00006292
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 240
Flash Point (°C): 107
Specific Gravity (20/20): 1.12
Other Product Names / Synonyms: Ethyl Picolinate, Picolinic Acid Ethyl Ester, Pyridine-2-carboxylic Acid Ethyl Ester
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Boiling Pt: 240 °C Density: 1.12 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00006292 CAS Number: 2524-52-9 EINECS: 219-758-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...