2-Pyridylacetonitrile ≥98.0%

Supplier: TCI America

Synonyms: 2-Pyridineacetonitrile

P1014-25G P1014-100G
TCP1014-025GEA 64 USD
TCP1014-025G TCP1014-100G
2-Pyridylacetonitrile ≥98.0%
2-Pyridylacetonitrile

CAS Number: 2739-97-1
MDL Number: MFCD00006346
Molecular Formula: C7H6N2
Molecular Weight: 118.14
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Flash Point (°C): 94
Freezing point (°C): 25


Other Product Names / Synonyms: 2-(Cyanomethyl)pyridine, 2-Pyridylacetonitrile

Formula: C₇H₆N₂
MW: 118.14
Flash Pt: 94 °C
MDL Number: MFCD00006346
CAS Number: 2739-97-1
EINECS: 220-364-6
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/16/51/28991651.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR