Neopentylbenzene ≥97.0%

Supplier: TCI America

Synonyms: (2,2-Dimethyl-1-propyl)benzene

P1013-5ML
TCP1013-005MLEA 148.19 USD
TCP1013-005ML
Neopentylbenzene ≥97.0%
Neopentylbenzene

CAS Number: 1007-26-7
MDL Number: MFCD00038279
Molecular Formula: C11H16
Molecular Weight: 148.25
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 186
Flash Point (°C): 50
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: (2,2-Dimethylpropyl)benzene

Formula: C₆H₅CH₂C((CH₃)₂)CH₃
MW: 148.25 g/mol
Boiling Pt: 186 °C
Density: 0.86 (20 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00038279
CAS Number: 1007-26-7
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR