Phenylmalonic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-phenylpropanedioic acid
P0987-25G
TCP0987-025GEA
34.53
USD
TCP0987-025G
Phenylmalonic acid ≥95.0% (by titrimetric analysis)
Phenylmalonic acid
CAS Number: 2613-89-0
MDL Number: MFCD00004253
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 153
Formula:
C₉H₈O₄ MW: 180.16 g/mol Melting Pt: 153 °C |
MDL Number:
MFCD00004253 CAS Number: 2613-89-0 EINECS: 220-044-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...