1-Phenylpiperazine ≥98.0%
Supplier: TCI America
Synonyms:
N-Phenylpiperazine
P0980-25G
P0980-250G
TCP0980-250GEA
424.32
USD
TCP0980-250G
TCP0980-25G
1-Phenylpiperazine ≥98.0%
1-Phenylpiperazine
CAS Number: 92-54-6
MDL Number: MFCD00005957
Molecular Formula: C10H14N2
Molecular Weight: 162.24
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 272
Flash Point (°C): 140
Freezing point (°C): 18
Specific Gravity (20/20): 1.07
Formula:
C₁₀H₁₄N₂ MW: 162.23 g/mol Boiling Pt: 272 °C Melting Pt: 17…19 °C Density: 1.07 (20 °C) Flash Pt: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005957 CAS Number: 92-54-6 EINECS: 202-165-6 UN: 2927 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...