(S)-(+)-Phenylsuccinic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(S)-(+)-Phenyl-1,4-butanedioic acid
P0936-1G
TCP0936-001GEA
163.37
USD
TCP0936-001G
(S)-(+)-Phenylsuccinic acid ≥98.0% (by titrimetric analysis)
(S)-(+)-Phenylsuccinic acid
CAS Number: 4036-30-0
MDL Number: MFCD00065929
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 174
Specific rotation [a]20/D: 147.5 deg (C=2, EtOH)
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Boiling Pt: 368 °C (1013 hPa) Melting Pt: 174 °C |
MDL Number:
MFCD00065929 CAS Number: 4036-30-0 EINECS: 223-719-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...