2,3,4,5,6-Pentafluoroaniline ≥98.0%

Supplier: TCI America
Warning

Synonyms: PentafluoroanilineAminopentafluorobenzene

P0922-25G
TCP0922-25GEA 240.43 USD
TCP0922-25G
2,3,4,5,6-Pentafluoroaniline ≥98.0%
2,3,4,5,6-Pentafluoroaniline

CAS Number: 771-60-8
MDL Number: MFCD00007643
Molecular Formula: C6H2F5N
Molecular Weight: 183.08
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 153
Flash Point (°C): 73
Freezing point (°C): 34

Formula: C₆H₂F₅N
MW: 183.08 g/mol
Boiling Pt: 153 °C
Melting Pt: 36…37 °C
Density: 1.744 g/cm³ (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007643
CAS Number: 771-60-8
EINECS: 212-234-2
https://us.vwr.com/stibo/web/std.lang.all/15/86/28991586.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR