4-Pentylphenol ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
4-Amylphenol
CAS Number: 14938-35-3
MDL Number: MFCD00020211
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 250
Flash Point (°C): 104
Freezing point (°C): 22
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Analytical Science, Alkylphenols (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: 4-Pentylphenol
Formula:
C₁₁H₁₆O MW: 164.25 g/mol Boiling Pt: 250 °C Melting Pt: 23 °C Density: 0.9597 g/cm³ (20 °C) Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020211 CAS Number: 14938-35-3 EINECS: 239-015-4 UN: 2430 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...