2-Propylvaleric acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Propylpentanoic acid Valproic Acid

P0823-100ML P0823-500ML P0823-25ML
TCP0823-100MLEA 121.71 USD
TCP0823-100ML TCP0823-500ML TCP0823-025ML
2-Propylvaleric acid ≥99.0% (by GC, titration analysis)
2-Propylvaleric acid

CAS Number: 99-66-1
MDL Number: MFCD00002672
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: 99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 126
Specific Gravity (20/20): 0.91


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents, Voltage-dependent Calcium Channel (VDCC) Antagonists, Epigenetics Research Reagents
Other Product Names / Synonyms: Dipropylacetic Acid, Valproic Acid, 2-Propylpentanoic Acid

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 220 °C
Density: 0.91 (20 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002672
CAS Number: 99-66-1
EINECS: 202-777-3
UN: 3265
ADR: 8,III
Merck Index: 12,10049
https://us.vwr.com/stibo/web/std.lang.all/14/93/28991493.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR