(Phenylthio)acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0753-250G P0753-25G
TCP0753-250GEA 532 USD
TCP0753-250G TCP0753-025G
(Phenylthio)acetic acid ≥98.0% (by titrimetric analysis)
(Phenylthio)acetic acid

CAS Number: 103-04-8
MDL Number: MFCD00004355
Molecular Formula: C8H8O2S
Molecular Weight: 168.21
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 64


Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: (Phenylmercapto)acetic Acid, S-Phenylthioglycolic Acid, Thiophenoxyacetic Acid

Formula: C₈H₈O₂S
Melting Pt: 64 °C
MDL Number: MFCD00004355
CAS Number: 103-04-8
EINECS: 203-073-9
https://us.vwr.com/stibo/web/std.lang.all/14/36/28991436.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR