D-Panthenol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dexpanthenol(R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

P0692-500G P0692-25G
TCP0692-025GEA 38.4 USD
TCP0692-025G TCP0692-500G
D-Panthenol ≥98.0% (by titrimetric analysis)
D-Panthenol

CAS Number: 81-13-0
MDL Number: MFCD00065006
Molecular Formula: C9H19NO4
Molecular Weight: 205.25
Purity/Analysis Method: 98.0% (T)
Form: Starch Syrup
Specific rotation [a]20/D: 31 deg (C=5, H2O)


Other Product Names / Synonyms: (R)-3-(2,4-Dihydroxy-3,3-dimethylbutyramido)-1-propanol, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, D-Pantothenyl Alcohol

Formula: C₉H₁₉NO₄
MW: 205.25 g/mol
MDL Number: MFCD00065006
CAS Number: 81-13-0
EINECS: 201-327-3
https://us.vwr.com/stibo/web/std.lang.all/13/98/28991398.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR