2-Pentyne ≥97.0%

Supplier: TCI America

P0628-25ML P0628-5ML
TCP0628-005MLEA 48.72 USD
TCP0628-005ML TCP0628-25ML
2-Pentyne ≥97.0%
2-Pentyne

CAS Number: 627-21-4
MDL Number: MFCD00009380
Molecular Formula: C5H8
Molecular Weight: 68.12
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 56
Flash Point (°C): -20
Specific Gravity (20/20): 0.71

Formula: C₅H₈
MW: 68.12 g/mol
Boiling Pt: 56 °C
Melting Pt: –109 °C
Density: 0.71 (20 °C)
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009380
CAS Number: 627-21-4
EINECS: 210-989-2
UN: 1993
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/13/53/28991353.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR