2-Pyrrolidone ≥98.0%
Supplier: TCI America
Synonyms:
2-Pyrrolidinone
, Pyrrolidone
P0575-25G
P0575-500G
TCP0575-500GEA
40.95
USD
TCP0575-500G
TCP0575-025G
2-Pyrrolidone ≥98.0%
2-Pyrrolidone
CAS Number: 616-45-5
MDL Number: MFCD00005270
Molecular Formula: C4H7NO
Molecular Weight: 85.11
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 245
Flash Point (°C): 145
Freezing point (°C): 24
Other Product Names / Synonyms: 2-Azacyclopentanone, gamma-Butyrolactam, 2-Ketopyrrolidine, 2-Pyrrolidinone
Formula:
C₄H₇NO MW: 85.11 g/mol Boiling Pt: 245 °C Melting Pt: 24…26 °C Density: 1.1115 g/cm³ (20 °C) Flash Pt: 145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005270 CAS Number: 616-45-5 EINECS: 210-483-1 Merck Index: 12,08200 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...