Pyridoxal hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride

P0559-1G P0559-25G P0559-5G
TCP0559-25GEA 165.39 USD
TCP0559-25G TCP0559-001G TCP0559-005G
Pyridoxal hydrochloride ≥98.0% (by HPLC, titration analysis)
Pyridoxal hydrochloride

CAS Number: 65-22-5
MDL Number: MFCD00012809
Molecular Formula: C8H9NO3
Molecular Weight: 203.62
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Formula: C₈H₁₀ClNO₃
MW: 203.62 g/mol
MDL Number: MFCD00012809
CAS Number: 65-22-5
EINECS: 200-602-5
https://us.vwr.com/stibo/web/std.lang.all/12/89/28991289.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR