Ethyl propiolate ≥98.0%

Supplier: TCI America

Synonyms: 2-Propynoic acid ethyl ester Propiolic acid ethyl ester Ethyl propynoate

P0529-5ML P0529-25ML
TCP0529-005MLEA 44.66 USD
TCP0529-005ML TCP0529-025ML
Ethyl propiolate ≥98.0%
Ethyl propiolate

CAS Number: 623-47-2
MDL Number: MFCD00009184
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Melting point (°C): 9
Flash Point (°C): 25
Specific Gravity (20/20): 0.96


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Propargylic Acid Ethyl Ester, Propiolic Acid Ethyl Ester, Ethyl Propargylate

Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 119 °C
Melting Pt: 9 °C
Density: 0.96 (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00009184
CAS Number: 623-47-2
EINECS: 210-795-8
UN: 3272
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/12/42/28991242.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR