2-Propanethiol ≥96.0%

Supplier: TCI America

Synonyms: Isopropyl mercaptanIsopropanethiol2-Mercaptopropane

P0489-500ML P0489-25ML
TCP0489-25MLEA 19.36 USD
TCP0489-25ML TCP0489-500ML
2-Propanethiol ≥96.0%
2-Propanethiol

CAS Number: 75-33-2
MDL Number: MFCD00004863
Molecular Formula: C3H8S
Molecular Weight: 76.16
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 53
Melting point (°C): -130
Flash Point (°C): -34
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Isopropyl Mercaptan

Formula: C₃H₈S
MW: 76.16 g/mol
Boiling Pt: 53 °C
Melting Pt: –130 °C
Density: 0.82 (20 °C)
Flash Pt: –34 °C
Storage Temperature: Ambient
MDL Number: MFCD00004863
CAS Number: 75-33-2
EINECS: 200-861-4
UN: 2402
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/11/78/28991178.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR