Propagyl bromide ≥97.0% stabilized
Supplier: TCI America
Synonyms:
3-Bromopropyne
Stabilization:
Stabilised with Stabilised
(stabilized with MgO)
CAS Number: 106-96-7
MDL Number: MFCD00000241
Molecular Formula: C3H3Br
Molecular Weight: 118.96
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 10
Freezing point (°C): -61
Specific Gravity (20/20): 1.58
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 3-Bromo-1-propyne
Formula:
C₃H₃Br Boiling Pt: 90 °C Melting Pt: –43 °C Density: 1.58 (20 °C) Flash Pt: 10 °C |
MDL Number:
MFCD00000241 CAS Number: 106-96-7 EINECS: 203-447-1 UN: 2345 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...