2-Methylpiperidine ≥98.0%

Supplier: TCI America

P0443-100ML P0443-25ML P0443-500ML
TCP0443-100MLEA 41 USD
TCP0443-100ML TCP0443-25ML TCP0443-500ML
2-Methylpiperidine ≥98.0%
2-Methylpiperidine

CAS Number: 109-05-7
MDL Number: MFCD00005982
Molecular Formula: C6H13N
Molecular Weight: 99.18
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 10
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: 2-Pipecoline

Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 118 °C
Melting Pt: –4.6 °C
Density: 0.84 (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00005982
CAS Number: 109-05-7
EINECS: 203-642-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR