(±)-Pipecolinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (±)-2-Piperidinecarboxylic acid H-DL-Homopro-OH

P0442-25G
TCP0442-025GEA 83.23 USD
TCP0442-025G
(±)-Pipecolinic acid ≥98.0% (by titrimetric analysis)
(±)-Pipecolinic acid

CAS Number: 535-75-1
MDL Number: MFCD00064347
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: DL-Homoproline, DL-Pipecolinic Acid, (+/-)-2-Piperidinecarboxylic Acid

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD00064347
CAS Number: 535-75-1
EINECS: 208-616-3
https://us.vwr.com/stibo/web/std.lang.all/11/31/28991131.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR