(1S)-(-)-α-Pinene ≥97.0%

Supplier: TCI America

Synonyms: (-)-Pin-2(3)-ene L(-)-α-Pinene (1S)-(-)-2-Pinene

P0440-500ML P0440-25ML
TCP0440-25MLEA 30.43 USD
TCP0440-25ML TCP0440-500ML
(1S)-(-)-α-Pinene ≥97.0%
(1S)-(-)-α-Pinene

CAS Number: 7785-26-4
MDL Number: MFCD00064145
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Melting point (°C): -64
Flash Point (°C): 32
Specific Gravity (20/20): 0.86
Specific rotation [a]20/D: -45.5 deg (neat)

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 155 °C
Melting Pt: –64 °C
Density: 0.86 (20 °C)
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064145
CAS Number: 7785-26-4
EINECS: 232-077-3
UN: 2368
ADR: 3,III
Merck Index: 12,07599
https://us.vwr.com/stibo/web/std.lang.all/11/26/28991126.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR