3-Picoline-N-oxide ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3-Picoline N-oxide3-Methylpyridine 1-oxide3-Methylpyridine-1-oxide3-Methylpyridine-N-oxide3-Picoline 1-oxide

P0419-25G
TCP0419-025GEA 152 USD
TCP0419-025G
3-Picoline-N-oxide ≥98.0%
3-Picoline N-oxide

CAS Number: 1003-73-2
MDL Number: MFCD00006203
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 37
Flash Point (°C): 143


Other Product Names / Synonyms: 3-Picoline N-Oxide

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 150 °C
Melting Pt: 37 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 143 °C
MDL Number: MFCD00006203
CAS Number: 1003-73-2
EINECS: 213-714-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR