3-Picoline ≥98.0%

Supplier: TCI America
Danger

Synonyms: β-Picoline3-Methylpyridine

P0416-25ML P0416-500ML
TCP0416-500MLEA 40.59 USD
TCP0416-500ML TCP0416-25ML
3-Picoline ≥98.0%
3-Picoline

CAS Number: 108-99-6
MDL Number: MFCD00006402
Molecular Formula: C6H7N
Molecular Weight: 93.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Melting point (°C): -18
Flash Point (°C): 36
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: 3-Picoline

Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 142 °C
Melting Pt: –18 °C
Density: 0.96 (20 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00006402
CAS Number: 108-99-6
EINECS: 203-636-9
UN: 2313
ADR: 3,III
Merck Index: 12,07555
https://us.vwr.com/stibo/web/std.lang.all/11/12/28991112.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR