Phthalhydrazide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,3-Dihydrophthalazine-1,4-dione

P0354-25G P0354-500G
TCP0354-025GEA 25.06 USD
TCP0354-025G TCP0354-500G
Phthalhydrazide ≥98.0% (by HPLC, titration analysis)
Phthalhydrazide

CAS Number: 1445-69-8
MDL Number: MFCD00006888
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2,3-Dihydro-1,4-phthalazinedione, 1,4-Phthalazinedione, Phthaloylhydrazine

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00006888
CAS Number: 1445-69-8
EINECS: 215-893-4
https://us.vwr.com/stibo/web/std.lang.all/10/81/28991081.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR