TCIAP0334

P0334-25ML
TCP0334-25MLEA 37.4 USD
TCP0334-25ML
TCIAP0334
CAS Number: 824-72-6
MDL Number: MFCD00002070
Molecular Formula: C6H5Cl2OP
Molecular Weight: 194.98
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 258
Flash Point (°C): 153
Freezing point (°C): 3
Specific Gravity (20/20): 1.40
Lambda max.: 267 nm (Cyclohexane)

Other Product Names / Synonyms: Benzenephosphonic Dichloride
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR