Dipropyl phthalate ≥98.0%

Supplier: TCI America

P0306-25ML P0306-100ML P0306-500ML
TCP0306-500MLEA 512.6 USD
TCP0306-500ML TCP0306-025ML TCP0306-100ML
Dipropyl phthalate ≥98.0%
Dipropyl phthalate

CAS Number: 131-16-8
MDL Number: MFCD00009371
Molecular Formula: C14H18O4
Molecular Weight: 250.29
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 305
Flash Point (°C): 110
Specific Gravity (20/20): 1.08


Product Categories: Materials Chemistry, Resin Additives, Analytical Science, Phthalates (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Phthalic Acid Dipropyl Ester

Formula: C₁₄H₁₈O₄
Boiling Pt: 305 °C
Density: 1.08 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009371
CAS Number: 131-16-8
EINECS: 205-015-8
UN: 3082
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/10/34/28991034.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR