Benzyl butyl phthalate ≥97.0%

Supplier: TCI America
Danger

Synonyms: BBP

P0288-25G P0288-500G
TCP0288-25GEA 28.4 USD
TCP0288-25G TCP0288-500G
Benzyl butyl phthalate ≥97.0%
Benzyl butyl phthalate

CAS Number: 85-68-7
MDL Number: MFCD00009440
Molecular Formula: C19H20O4
Molecular Weight: 312.37
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 250
Flash Point (°C): 213
Specific Gravity (20/20): 1.12


Product Categories: Materials Chemistry, Resin Additives, Analytical Science, Phthalates (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Phthalic Acid Benzyl Butyl Ester, BBP

Formula: C₁₉H₂₀O₄
MW: 312.37 g/mol
Boiling Pt: 250 °C
Melting Pt: –35 °C
Density: 1.12 (20 °C)
Flash Pt: 213 °C
Storage Temperature: Ambient
MDL Number: MFCD00009440
CAS Number: 85-68-7
EINECS: 201-622-7
UN: 3082
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/10/05/28991005.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR