Phthalamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

P0283-500G P0283-25G
TCP0283-500GEA 368 USD
TCP0283-500G TCP0283-025G
Phthalamide ≥98.0% (by total nitrogen basis)
Phthalamide

CAS Number: 88-96-0
MDL Number: MFCD00025478
Molecular Formula: C8H8N2O2
Molecular Weight: 164.16
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 223

Formula: C₈H₈N₂O₂
Melting Pt: 223 °C
MDL Number: MFCD00025478
CAS Number: 88-96-0
EINECS: 201-870-6
https://us.vwr.com/stibo/web/std.lang.all/10/00/28991000.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR