Trichloro(phenyl)silane ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Phenyltrichlorosilane
P0240-25G
P0240-500G
TCP0240-500GEA
110.64
USD
TCP0240-500G
TCP0240-25G
Trichloro(phenyl)silane ≥98.0%
Trichloro(phenyl)silane
CAS Number: 98-13-5
MDL Number: MFCD00000479
Molecular Formula: C6H5Cl3Si
Molecular Weight: 211.54
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 91
Specific Gravity (20/20): 1.32
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Trichlorophenylsilane
Formula:
C₆H₅Cl₃Si MW: 211.55 g/mol Boiling Pt: 201 °C Melting Pt: –40 °C Density: 1.32 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00000479 CAS Number: 98-13-5 EINECS: 202-640-8 UN: 1804 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...