Phenylhydrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

P0183-25G P0183-500G P0183-100G
TCP0183-25GEA 34.35 USD
TCP0183-25G TCP0183-100G TCP0183-500G
Phenylhydrazine ≥98.0% (by GC, titration analysis)
Phenylhydrazine

CAS Number: 100-63-0
MDL Number: MFCD00007573
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 88
Freezing point (°C): 19
Specific Gravity (20/20): 1.10

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 19.6 °C
Density: 1.10 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00007573
CAS Number: 100-63-0
EINECS: 202-873-5
UN: 2572
ADR: 6.1,II
Merck Index: 12,07447
https://us.vwr.com/stibo/web/std.lang.all/09/19/28990919.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR