4-Phenylazophenol ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-(Phenyldiazenyl)phenol

P0149-25G
TCP0149-025GEA 46.67 USD
TCP0149-025G
4-Phenylazophenol ≥95.0% (by titrimetric analysis)
4-Phenylazophenol

CAS Number: 1689-82-3
MDL Number: MFCD00002330
Molecular Formula: C12H10N2O
Molecular Weight: 198.23
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Boiling point (°C): 230
Melting point (°C): 153


Other Product Names / Synonyms: 4-Hydroxyazobenzene, Solvent Yellow 7

Formula: C₁₂H₁₀N₂O
MW: 198.23 g/mol
Boiling Pt: 230 °C
Melting Pt: 153 °C
MDL Number: MFCD00002330
CAS Number: 1689-82-3
EINECS: 216-880-6
https://us.vwr.com/stibo/web/std.lang.all/08/68/28990868.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR