Phenothiazine ≥98.0%

Supplier: TCI America
Warning

Synonyms: 10H-Phenothiazine

P0106-25G P0106-500G
TCP0106-025GEA 27.94 USD
TCP0106-025G TCP0106-500G
Phenothiazine ≥98.0%
Phenothiazine

CAS Number: 92-84-2
MDL Number: MFCD00005015
Molecular Formula: C12H9NS
Molecular Weight: 199.27
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 183
Flash Point (°C): 202


Product Categories: Materials Chemistry, Polymerization Inhibitors
Other Product Names / Synonyms: 2,3:5,6-Dibenzo-1,4-thiazine, Thiodiphenylamine

Formula: C₁₂H₉NS
MW: 199.28 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 183 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: 202 °C
MDL Number: MFCD00005015
CAS Number: 92-84-2
EINECS: 202-196-5
Merck Index: 12,07404
https://us.vwr.com/stibo/web/std.lang.all/08/02/28990802.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR