Phenazine ≥99.0% (by HPLC)

Supplier: TCI America

P0082-1G P0082-25G P0082-5G
TCP0082-025GEA 129.92 USD
TCP0082-025G TCP0082-5G TCP0082-001G
Phenazine ≥99.0% (by HPLC)
Phenazine

CAS Number: 92-82-0
MDL Number: MFCD00005023
Molecular Formula: C12H8N2
Molecular Weight: 180.21
Purity/Analysis Method: 99.0% (HPLC)
Form: Crystal
Melting point (°C): 176


Other Product Names / Synonyms: Azophenylene

Formula: C₁₂H₈N₂
MW: 180.21 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.26 g/cm³ (22 °C)
MDL Number: MFCD00005023
CAS Number: 92-82-0
EINECS: 202-193-9
Merck Index: 12,07359
https://us.vwr.com/stibo/web/std.lang.all/07/80/28990780.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR