1-Pentyne ≥98.0%
Supplier: TCI America
P0068-25ML
TCP0068-025MLEA
69.96
USD
TCP0068-025ML
1-Pentyne ≥98.0%
1-Pentyne
CAS Number: 627-19-0
MDL Number: MFCD00009469
Molecular Formula: C5H8
Molecular Weight: 68.12
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 40
Flash Point (°C): -34
Specific Gravity (20/20): 0.70
Formula:
C₅H₈ MW: 68.12 g/mol Boiling Pt: 40 °C Melting Pt: –90 °C Density: 0.70 (20 °C) Flash Pt: –34 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009469 CAS Number: 627-19-0 EINECS: 210-987-1 UN: 1993 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...