2-Pentene ≥95.0%

Supplier: TCI America

P0064-25ML
TCP0064-025MLEA 46.8 USD
TCP0064-025ML
2-Pentene ≥95.0%
2-Pentene

CAS Number: 109-68-2
MDL Number: MFCD00009384
Molecular Formula: C5H10
Molecular Weight: 70.14
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 36
Flash Point (°C): -45
Specific Gravity (20/20): 0.65


Other Product Names / Synonyms: 2-Amylene

Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 36 °C
Melting Pt: –140 °C
Density: 0.65 (20 °C)
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00009384
CAS Number: 109-68-2
EINECS: 203-695-0
UN: 3295
ADR: 3,II
Merck Index: 12,07263
https://us.vwr.com/stibo/web/std.lang.all/07/70/28990770.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR