3-Pentanone ≥98.0%

Supplier: TCI America
Danger

Synonyms: Diethyl ketone

P0061-500ML P0061-25ML
TCP0061-25MLEA 28.23 USD
TCP0061-25ML TCP0061-500ML
3-Pentanone ≥98.0%
3-Pentanone

CAS Number: 96-22-0
MDL Number: MFCD00009320
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 13
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Diethyl Ketone, Propione

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 102 °C
Melting Pt: –40 °C
Density: 0.82 (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009320
CAS Number: 96-22-0
EINECS: 202-490-3
UN: 1156
ADR: 3,II
Merck Index: 13,03148
https://us.vwr.com/stibo/web/std.lang.all/07/68/28990768.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR