2-Methyl-2-butanol ≥98.0%

Supplier: TCI America
Danger

Synonyms: tert-Amyl alcoholtert-Pentyl alcoholtert-Pentanol

P0059-25ML P0059-500ML
TCP0059-500MLEA 67.5 USD
TCP0059-500ML TCP0059-25ML
2-Methyl-2-butanol ≥98.0%
2-Methyl-2-butanol

CAS Number: 75-85-4
MDL Number: MFCD00004478
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Melting point (°C): -8
Flash Point (°C): 41
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Ethyldimethylcarbinol, 2-Methyl-2-butanol, tert-Pentanol

Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 102 °C
Melting Pt: –8 °C
Density: 0.81 (20 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00004478
CAS Number: 75-85-4
EINECS: 200-908-9
UN: 1105
ADR: 3,II
Merck Index: 12,07282
https://us.vwr.com/stibo/web/std.lang.all/07/66/28990766.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR