(±)-2-Pentanol ≥98.0%

Supplier: TCI America
Danger

Synonyms: (±)-Methyl propyl carbinolDL-2-Pentanol(±)-Propyl methyl carbinol(±)-2-Hydroxy pentane

P0056-500ML P0056-25ML
TCP0056-500MLEA 142.44 USD
TCP0056-500ML TCP0056-25ML
(±)-2-Pentanol ≥98.0%
(±)-2-Pentanol

CAS Number: 6032-29-7
MDL Number: MFCD00004579
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 34
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Methylpropylcarbinol

Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 118 °C
Melting Pt: –50 °C
Density: 0.81 (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00004579
CAS Number: 6032-29-7
EINECS: 227-907-6
UN: 1105
ADR: 3,III
Merck Index: 12,07258
https://us.vwr.com/stibo/web/std.lang.all/07/64/28990764.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR